Order Entry
Netherlands
ContactUsLinkComponent
 
1-Cyclopropyl-3-[(4-fluorophenyl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one, tech
Catalog # APOSPC28191-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Cyclopropyl-3-[(4-fluorophenyl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one, tech
Catalog # APOSPC28191-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.276 g/mol
Storage Temperature:Ambient
CAS Number:478247-85-7
Formula:C₁₅H₁₄FNO₂
MDL Number:MFCD01871749