(E)-N'-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}-N,N-dimethylmethanimidamide, tech
Catalog # APOSPC28168-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:266.651 g/mol
Storage Temperature:Ambient
CAS Number:339029-44-6
Formula:C₉H₁₀ClF₃N₄
MDL Number:MFCD00215321