N-[2-(4-Methylpiperazin-1-yl)-5-(trifluoromethyl)phenyl]hydroxylamine, tech
Catalog # APOSPC28156-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:275.27 g/mol
Storage Temperature:Ambient
CAS Number:866133-93-9
Formula:C₁₂H₁₆F₃N₃O
MDL Number:MFCD04126038