Order Entry
Netherlands
ContactUsLinkComponent
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]ethan-1-ol, tech
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]ethan-1-ol, tech
Catalog # APOSPC28151-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]ethan-1-ol, tech
Catalog # APOSPC28151-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :230.186 g/mol
Storage Temperature:Ambient
CAS Number:116040-98-3
Formula:C₁₀H₉F₃N₂O
MDL Number:MFCD01017153