2-Chloro-2,2-difluoro-1-(1-methyl-1H-indol-3-yl)ethan-1-ol, tech
Catalog # APOSPC28150-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:245.653 g/mol
Storage Temperature:Ambient
CAS Number:478079-32-2
Formula:C₁₁H₁₀ClF₂NO
MDL Number:MFCD02571244