(2R)-3-{1,4-Dioxa-8-azaspiro[4.5]decan-8-yl}-1,1,1-trifluoropropan-2-ol, tech
Catalog # APOSPC28149-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:255.234 g/mol
Storage Temperature:Ambient
CAS Number:477762-19-9
Formula:C₁₀H₁₆F₃NO₃
MDL Number:MFCD06809966