2-Chloro-2,2-difluoro-1-(1H-indol-3-yl)ethan-1-ol, tech
Catalog # APOSPC28147-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:231.626 g/mol
Storage Temperature:Ambient
CAS Number:478079-28-6
Formula:C₁₀H₈ClF₂NO
MDL Number:MFCD02571237