1-[2-(3-Fluorophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-one 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:879626-93-4
:
Formula:C₁₂H₁₀FNOS
MDL Number:MFCD09304610
:235.277 g/mol
Storage Temperature:Ambient
Frequently Bought Together