Order Entry
Netherlands
ContactUsLinkComponent
(3Z)-4-(2,3-Dihydro-1H-indol-1-yl)-1,1-difluorobut-3-en-2-one, tech
(3Z)-4-(2,3-Dihydro-1H-indol-1-yl)-1,1-difluorobut-3-en-2-one, tech
Catalog # APOSPC28051-1G
Supplier:  Apollo Scientific
CAS Number:  
(3Z)-4-(2,3-Dihydro-1H-indol-1-yl)-1,1-difluorobut-3-en-2-one, tech
Catalog # APOSPC28051-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :223.219 g/mol
Storage Temperature:Ambient
CAS Number:478040-92-5
Formula:C₁₂H₁₁F₂NO
MDL Number:MFCD02186227