Order Entry
Netherlands
ContactUsLinkComponent
 
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]propanoic acid, tech
Catalog # APOSPC28034-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]propanoic acid, tech
Catalog # APOSPC28034-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :258.197 g/mol
Storage Temperature:Ambient
CAS Number:478030-59-0
Formula:C₁₁H₉F₃N₂O₂
MDL Number:MFCD02571586