Order Entry
Netherlands
ContactUsLinkComponent
3-(2-Fluorophenyl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech
3-(2-Fluorophenyl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech
Catalog # APOSPC28019-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(2-Fluorophenyl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech
Catalog # APOSPC28019-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :299.296 g/mol
Storage Temperature:Ambient
CAS Number:383148-31-0
Formula:C₁₇H₁₄FNO₃
MDL Number:MFCD03011837