Order Entry
Netherlands
ContactUsLinkComponent
(3E)-3-[2-(4-Fluorophenyl)hydrazin-1-ylidene]-1-methyl-2,3-dihydro-1H-indol-2-one, tech
(3E)-3-[2-(4-Fluorophenyl)hydrazin-1-ylidene]-1-methyl-2,3-dihydro-1H-indol-2-one, tech
Catalog # APOSPC27988-1G
Supplier:  Apollo Scientific
CAS Number:  
(3E)-3-[2-(4-Fluorophenyl)hydrazin-1-ylidene]-1-methyl-2,3-dihydro-1H-indol-2-one, tech
Catalog # APOSPC27988-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :269.274 g/mol
Storage Temperature:Ambient
CAS Number:320422-06-8
Formula:C₁₅H₁₂FN₃O
MDL Number:MFCD01314820