Order Entry
Netherlands
ContactUsLinkComponent
[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methyl 2-chloroacetate, tech
[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methyl 2-chloroacetate, tech
Catalog # APOSPC27969-1G
Supplier:  Apollo Scientific
CAS Number:  
[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methyl 2-chloroacetate, tech
Catalog # APOSPC27969-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :291.055 g/mol
Storage Temperature:Ambient
CAS Number:303148-55-2
Formula:C₈H₇Cl₂F₃N₂O₂
MDL Number:MFCD00141485