Order Entry
Netherlands
ContactUsLinkComponent
6-(Trifluoromethyl)pyrazin-2-amine 97%
6-(Trifluoromethyl)pyrazin-2-amine 97%
Catalog # APOSPC201282-250MG
Supplier:  Apollo Scientific
CAS Number:  
6-(Trifluoromethyl)pyrazin-2-amine 97%
Catalog # APOSPC201282-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :163.101 g/mol
Storage Temperature:Ambient
CAS Number:69816-35-9
Formula:C₅H₄F₃N₃
MDL Number:MFCD10697803