Order Entry
Netherlands
ContactUsLinkComponent
(6-(Trifluoromethyl)pyridin-2-yl)methanol 98%
(6-(Trifluoromethyl)pyridin-2-yl)methanol 98%
Catalog # APOSPC201117-1G
Supplier:  Apollo Scientific
CAS Number:  
(6-(Trifluoromethyl)pyridin-2-yl)methanol 98%
Catalog # APOSPC201117-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.124 g/mol
Storage Temperature:Ambient
CAS Number:131747-53-0
Formula:C₇H₆F₃NO
MDL Number:MFCD11100723