Order Entry
Netherlands
ContactUsLinkComponent
Methyl 4-(chloromethyl)-3,5-difluorobenzoate 97%
Methyl 4-(chloromethyl)-3,5-difluorobenzoate 97%
Catalog # APOSPC201105-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl 4-(chloromethyl)-3,5-difluorobenzoate 97%
Catalog # APOSPC201105-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :220.6 g/mol
Storage Temperature:Ambient
CAS Number:1263283-69-7
Formula:C₉H₇ClF₂O₂
MDL Number:MFCD17676601