0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Netherlands
ContactUsLinkComponent
5-(4-Fluorobenzyl)-1,3,4-thiadiazol-2-amine
5-(4-Fluorobenzyl)-1,3,4-thiadiazol-2-amine
5-(4-Fluorobenzyl)-1,3,4-thiadiazol-2-amine
APOSPC200519-1G
:
Apollo Scientific
:
5-(4-Fluorobenzyl)-1,3,4-thiadiazol-2-amine
APOSPC200519-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
209.243 g/mol
:
Ambient
:
39181-55-0
:
C₉H₈FN₃S
:
MFCD01364163