Order Entry
Netherlands
ContactUsLinkComponent
4-(4-Methoxyphenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-amine
4-(4-Methoxyphenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-amine
Catalog # APOSPC200450-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(4-Methoxyphenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-amine
Catalog # APOSPC200450-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :271.238 g/mol
Storage Temperature:Ambient
CAS Number:164668-12-6
Formula:C₁₂H₁₂F₃N₃O
MDL Number:MFCD12653214