Order Entry
Netherlands
ContactUsLinkComponent
 
3-Cyano-6-methyl-4-(trifluoromethyl)pyridine-2-carboxylic acid
Catalog # APOSPC200444-500MG
Supplier:  Apollo Scientific
CAS Number:  
3-Cyano-6-methyl-4-(trifluoromethyl)pyridine-2-carboxylic acid
Catalog # APOSPC200444-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :230.143 g/mol
Melting Pt:160 °C (dec.)
Storage Temperature:Ambient
CAS Number:1565827-83-9
Formula:C₉H₅F₃N₂O₂
MDL Number:MFCD27957326