Order Entry
Netherlands
ContactUsLinkComponent
Potassium {4-[(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)methyl]phenyl}trifluoroboranuide
Potassium {4-[(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)methyl]phenyl}trifluoroboranuide
Catalog # APOSPC200438-5G
Supplier:  Apollo Scientific
CAS Number:  
Potassium {4-[(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)methyl]phenyl}trifluoroboranuide
Catalog # APOSPC200438-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :343.15 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₀BF₃NO₂·K
MDL Number:MFCD27957313