Methyl 4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzoate
Catalog # APOSPC200431-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:297.229 g/mol
Melting Pt:76…78 °C
Storage Temperature:Ambient
CAS Number:869109-13-7
Formula:C₁₄H₁₀F₃NO₃
MDL Number:MFCD27957294