Methyl 3-bromo-4-(((trifluoromethyl)sulphonyl)oxy)-5-(trimethylsilyl)benzoate
Catalog # APOSPC200120-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Verpakking:Bottle
- VPE:100 mg
- Seq:2440
Specifications
About this item
:
:435.286 g/mol
Storage Temperature:Kamertemperatuur
UN:1759
ADR:8,III
CAS Number:1242069-33-5
Formula:C₁₂H₁₄BrF₃O₅SSi
MDL Number:MFCD25948970