Methyl 3-bromo-5-(((trifluoromethyl)sulphonyl)oxy)-4-(trimethylsilyl)benzoate
Catalog # APOSPC200118-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:435.286 g/mol
Storage Temperature:Ambient
CAS Number:1242069-32-4
Formula:C₁₂H₁₄BrF₃O₅SSi
MDL Number:MFCD25948969