

2,4-Dimethylfluorobenzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:452-65-3
:
Formula:C₈H₉F
MDL Number:MFCD00060643
:124.155 g/mol
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together