
1-Chloro-1,1-dibromo-2,2,2-trifluoroethane (FC-113aB2) 97%
Catalog # APOSPC1720-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:276.278 g/mol
Boiling Pt:91…92 °C
Melting Pt:42…45 °C
:GHS07
Storage Temperature:Ambient
CAS Number:754-17-6
Formula:C₂Br₂ClF₃
MDL Number:MFCD00042121