Order Entry
Netherlands
ContactUsLinkComponent
3'-Fluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
Irritant
3'-Fluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
Catalog # APOSPC11149-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3'-Fluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
Catalog # APOSPC11149-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :232.207 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:22494-43-5
Formula:C₁₃H₉FO₃
MDL Number:MFCD04039228