Order Entry
Netherlands
ContactUsLinkComponent
Ethyl 8-fluoro-4-hydroxy-6-iodoquinoline-3-carboxylate ≥98%
Ethyl 8-fluoro-4-hydroxy-6-iodoquinoline-3-carboxylate ≥98%
Catalog # APOSPC110107-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 8-fluoro-4-hydroxy-6-iodoquinoline-3-carboxylate ≥98%
Catalog # APOSPC110107-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :361.108 g/mol
Storage Temperature:Refrigerator
CAS Number:228728-08-3
Formula:C₁₂H₉FINO₃
MDL Number:MFCD12922926