Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(4-Fluorophenyl)-3-hydroxy-1H-1,2,4-triazole
1-(4-Fluorophenyl)-3-hydroxy-1H-1,2,4-triazole
1-(4-Fluorophenyl)-3-hydroxy-1H-1,2,4-triazole
APOSPC0926-250MG
:
Apollo Scientific
:
1-(4-Fluorophenyl)-3-hydroxy-1H-1,2,4-triazole
APOSPC0926-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
179.151 g/mol
:
GHS07
:
Ambient
:
42158-50-9
:
C₈H₆FN₃O
:
MFCD04035653