Order Entry
Netherlands
ContactUsLinkComponent
(1S)-1-(2-Fluorophenyl)ethan-1-ol
Irritant
(1S)-1-(2-Fluorophenyl)ethan-1-ol
Catalog # APOSPC0605-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(1S)-1-(2-Fluorophenyl)ethan-1-ol
Catalog # APOSPC0605-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :140.155 g/mol
Boiling Pt:110 °C ( 135 mm Hg)
Density:1.1 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:171032-87-4
Formula:C₈H₉FO
MDL Number:MFCD03092992