2-Hydroxy-5-(trifluoromethoxy)phenacyl bromide
Catalog # APOSPC0594-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:299.041 g/mol
Melting Pt:43…45 °C
Storage Temperature:Refrigerator
UN:1760
ADR:8,III
CAS Number:496052-53-0
Formula:C₉H₆BrF₃O₃
MDL Number:MFCD07366522