
4-Methyl-2-(trifluoromethyl)benzyl bromide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:261952-18-5
:
Formula:C₉H₈BrF₃
MDL Number:MFCD01631611
:253.059 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together