Order Entry
Netherlands
ContactUsLinkComponent
4-[4-(Hydroxymethyl)benzyl]piperazine, N1-BOC protected 95%
Corrosive
Irritant
4-[4-(Hydroxymethyl)benzyl]piperazine, N1-BOC protected 95%
Catalog # APOSOR9695-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
4-[4-(Hydroxymethyl)benzyl]piperazine, N1-BOC protected 95%
Catalog # APOSOR9695-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :306.4 g/mol
Melting Pt:52…53 °C
Storage Temperature:Ambient
UN:2923
ADR:8+6.1,III
 :GHS05
 :GHS07
CAS Number:622381-67-3
Formula:C₁₇H₂₆N₂O₃
MDL Number:MFCD06658975