tert-Butyl 3-(2-hydroxyethyl)piperidine-1-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:146667-84-7
:
Formula:C₁₂H₂₃NO₃
MDL Number:MFCD04114970
:229.316 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together