Order Entry
Netherlands
ContactUsLinkComponent
3-(4-Chlorophenyl)azetidine hydrochloride 95%
Irritant
3-(4-Chlorophenyl)azetidine hydrochloride 95%
Catalog # APOSOR964271-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(4-Chlorophenyl)azetidine hydrochloride 95%
Catalog # APOSOR964271-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :204.096 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:7606-31-7
Formula:C₉H₁₀ClN·ClH
MDL Number:MFCD08460697