Order Entry
Netherlands
ContactUsLinkComponent
3-Hydroxy-N-methoxy-N-methylbenzamide 95%
3-Hydroxy-N-methoxy-N-methylbenzamide 95%
Catalog # APOSOR957682-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Hydroxy-N-methoxy-N-methylbenzamide 95%
Catalog # APOSOR957682-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :181.189 g/mol
Storage Temperature:Ambient
CAS Number:615558-48-0
Formula:C₉H₁₁NO₃
MDL Number:MFCD18843718