Order Entry
Netherlands
ContactUsLinkComponent
Propyl 3-chlorobenzoate 95%
Propyl 3-chlorobenzoate 95%
Catalog # APOSOR957102-1G
Supplier:  Apollo Scientific
CAS Number:  
Propyl 3-chlorobenzoate 95%
Catalog # APOSOR957102-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198.646 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₁ClO₂
MDL Number:MFCD00457373