Order Entry
Netherlands
ContactUsLinkComponent
1-Cyclopentyl-5-nitro-1H-pyrazole 95%
1-Cyclopentyl-5-nitro-1H-pyrazole 95%
Catalog # APOSOR956274-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Cyclopentyl-5-nitro-1H-pyrazole 95%
Catalog # APOSOR956274-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :181.192 g/mol
Storage Temperature:Ambient
CAS Number:1170836-25-5
Formula:C₈H₁₁N₃O₂
MDL Number:MFCD09701860