Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl homovanillate
Methyl homovanillate 98%
Methyl homovanillate 98%
Methyl homovanillate 98%
APOSOR955876-1G
:
Apollo Scientific
:
Methyl homovanillate 98%
APOSOR955876-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
196.2 g/mol
:
Ambient
:
15964-80-4
:
C₁₀H₁₂O₄
:
MFCD00127727