(1S,3R)-Methyl 3-aminocyclopentanecarboxylate hydrochloride 96%
Catalog # APOSOR955812-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:179.645 g/mol
Storage Temperature:Ambient
CAS Number:180323-49-3
Formula:C₇H₁₃NO₂·ClH
MDL Number:MFCD23106006