Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Hydroxycinnamic acid methyl ester
3-Hydroxycinnamic acid methyl ester 95%
3-Hydroxycinnamic acid methyl ester 95%
APOSOR955453-1G
:
Apollo Scientific
:
3-Hydroxycinnamic acid methyl ester 95%
APOSOR955453-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
178.185 g/mol
:
Ambient
:
66417-46-7
:
C₁₀H₁₀O₃
:
MFCD09259949