
2-Chloro-3-methylisonicotinaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:790696-96-7
:
Formula:C₇H₆ClNO
MDL Number:MFCD09910248
:155.582 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together