7-Methyl-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:422318-36-3
:
Formula:C₁₀H₁₄N₂
MDL Number:MFCD07369975
:162.232 g/mol
Storage Temperature:Ambient
Frequently Bought Together