Order Entry
Netherlands
ContactUsLinkComponent
2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-1-carboxylic acid
Irritant
2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-1-carboxylic acid
Catalog # APOSOR948823-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-1-carboxylic acid
Catalog # APOSOR948823-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :227.257 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1251004-87-1
Formula:C₁₁H₁₇NO₄
MDL Number:MFCD17016465