Methyl (1R,2S,5S)-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylate hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:565456-77-1
:
Formula:C₉H₁₅NO₂·ClH
MDL Number:MFCD11865137
:205.682 g/mol
Storage Temperature:Ambient
Frequently Bought Together