Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(4-Bromophenoxy)benzonitrile
2-(4-Bromophenoxy)benzonitrile 98%
2-(4-Bromophenoxy)benzonitrile 98%
APOSOR946471-1G
:
Apollo Scientific
:
2-(4-Bromophenoxy)benzonitrile 98%
APOSOR946471-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
274.113 g/mol
:
GHS07
:
Ambient
:
330793-13-0
:
C₁₃H₈BrNO
:
MFCD10564769