tert-Butyl 2-(bromomethyl)azetidine-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1363380-84-0
:
Formula:C₉H₁₆BrNO₂
MDL Number:MFCD22566236
:250.133 g/mol
Storage Temperature:Ambient
Frequently Bought Together