tert-Butyl 2-azaspiro[3.4]oct-6-ene-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1638768-92-9
:
Formula:C₁₂H₁₉NO₂
MDL Number:MFCD27986984
:209.285 g/mol
Storage Temperature:Ambient
Frequently Bought Together