cis-5-Methyl-1H-hexahydropyrrolo[3,4-b]pyrrole dihydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1363166-00-0
:
Formula:C₇H₁₄N₂·2ClH
MDL Number:MFCD21364419
:199.121 g/mol
Storage Temperature:Ambient
Frequently Bought Together