Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyclopropyl(3-pyridinyl)methanone
Cyclopropyl(3-pyridinyl)methanone 95%
Cyclopropyl(3-pyridinyl)methanone 95%
APOSOR929114-1G
:
Apollo Scientific
:
Cyclopropyl(3-pyridinyl)methanone 95%
APOSOR929114-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
147.174 g/mol
:
Ambient
:
2811
:
6.1,III
:
24966-13-0
:
C₉H₉NO
:
MFCD09864467