Order Entry
Netherlands
ContactUsLinkComponent
2-(1H-Imidazol-1-yl)aniline 95%
2-(1H-Imidazol-1-yl)aniline 95%
Catalog # APOSOR927017-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(1H-Imidazol-1-yl)aniline 95%
Catalog # APOSOR927017-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :159.188 g/mol
Storage Temperature:Ambient
CAS Number:26286-54-4
Formula:C₉H₉N₃
MDL Number:MFCD03274531